Learn More
Akt1/PKBα Rabbit anti-Human, Mouse, Rat, Clone: AW24, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma 05796
Catalog No. 05-796-MI
Add to cart
Description
Description
Detect Akt1/PKBα using this Anti-Akt1 Antibody, clone AW24 validated for use in immunoprecipitation and western blotting.
Specifications
Specifications
Akt1/PKBα
AW24
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Peptide (C-RPHFPQFSYSASGTA) corresponding to the C-terminal (residues 466-480) of rat Akt1/PKBα
Protein A purfied
−20°C for 24 months
Monoclonal
Human, Murine, Rat
Immunoprecipitation, Western Blot
Unconjugated
NM_005163
Rabbit
IgG
100μL
Primary
AKT1, RAC, PRKBA, RAC-ALPHA, PKB, MGC99656, RAC-PK-alpha, C-AKT, AKT