Learn More
MilliporeSigma™ anti-Akt1/PKBα, Clone: AW24, ™ (Upstate™) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ 05796/DEL
Catalog No. 50-171-833
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Immunoprecipitation, Western Blotting
Abbreviations: EIA=ELISA, FC=Flow Cytometry, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, WB=Western Blotting,GS=Gel Supershift, EM=Electron Microscopy, CM=Confocal Microscopy, RIA=Radioimmunossay, AP=Affinity Purification Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
Akt1/PKBα
Semi-purified
IgG
RUO
Primary
Human, Murine, Rat
Unconjugated
Rabbit
100μL
Cell Signaling
Monoclonal