Learn More
AKT2, Mouse, Clone: 1B3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant AKT2.
Supplier: Abnova Corporation H00000208M01
Description
This gene is a putative oncogene encoding a protein belonging to a subfamily of serine/threonine kinases containing SH2-like (Src homology 2-like) domains. The gene was shown to be amplified and overexpressed in 2 of 8 ovarian carcinoma cell lines and 2 of 15 primary ovarian tumors. Overexpression contributes to the malignant phenotype of a subset of human ductal pancreatic cancers. The encoded protein is a general protein kinase capable of phophorylating several known proteins. [provided by RefSeq
Sequence: MRAIQMVANSLKQRAPGEDPMDYKCGSPSDSSTTEEMEVAVSKARAKVTMNDFDYLKLLGKGTFGKVILVREKATGRYYAMKILRKEVIISpecifications
AKT2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
M95936 | |
AKT2 | |
AKT2 (AAA58364, 100 a.a. ∼ 189 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunofluorescence, Western Blot | |
1B3 | |
Mouse monoclonal antibody raised against a partial recombinant AKT2. | |
AKT2 | |
PKBB/PKBBETA/PRKBB/RAC-BETA | |
Mouse | |
Affinity chromatography | |
RUO | |
208 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.