Learn More
Akt2/PKBΒ Rabbit anti-Human, Mouse, Rat, Clone: AW114, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier:  MilliporeSigma 04771
Catalog No. 04-771-MI
Add to Cart
Description
Description
Anti-Akt2/PKBβ Antibody, clone AW114 detects level of Akt2/PKBβ and has been published and validated for use in western blotting and immunoprecipitation.
Specifications
Specifications
Akt2/PKBβ
Monoclonal
Unconjugated
NP_001617.1
KLH-conjugated, synthetic peptide (CRYDSLGSLELDQRTH) corresponding to amino acids 455-469 of human Akt2/PKBβ, with an N-terminal cysteine added for conjugation purposes.
Primary
Human, Mouse, Rat
Purified
Immunoprecipitation, Western Blot
AW114
Cultured supernantant containing 0.05% sodium azide.
Rabbit
100 μL
EC 2.7.11.1, PKBB, PKBBETA, PRKBB, RAC-BETA, RAC-PK-beta
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
IgG