Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Akt2, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP126594

Catalog No. NBP126594

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

AKT2
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 5-10 ug/mg lysate
NP_001617.
EC 2.7.11, EC 2.7.11.1, Murine thymoma viral (v-akt) homolog-2, PKB beta, PKBB, PKBBETA, PRKBB, Protein kinase Akt-2, Protein kinase B beta, rac protein kinase beta, RAC-BETA, RAC-beta serine/threonine-protein kinase, RAC-PK-beta, v-akt murine thymoma viral oncogene homolog 2
Rabbit
IgG
0.1 ml
Cancer, mTOR Pathway
Primary
208
Western Blot, Immunoprecipitation
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
Affinity Purified
AKT2
The immunogen recognized by this antibody maps to a region between residue 431 and 481 of human v-akt murine thymoma viral oncogene homolog 2 using the numbering given in entry NP_001617.1 (GeneID 208).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only