Learn More
Akt3/PKBγ Mouse anti-Human, Clone: GMA104, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05780
Catalog No. 05-780-MI
Add to cart
Description
Description
This Anti-Akt3/PKBγ Antibody, clone GMA104 is validated for use in immunoprecipitation, western blotting for the detection of Akt3/PKBγ.
Specifications
Specifications
Akt3/PKBγ
GMA104
Buffered solution containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4.
Purified
Murine
IgG2a
100μg
Primary
Human
Immunoprecipitation, Western Blot
Unconjugated
NM_005465
AKT3, PRKBG, DKFZP434N0250, PKBG, RAC-gamma, RAC-PK-gamma, STK-2
peptide corresponding to amino acids 119-131 (CSPTSQIDNIGEE) of human Akt3/PKBγ
Protein A purfied
2-8°C for one year
Monoclonal