Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALS2CR11 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524265
Description
Specifications
ALS2CR11 | |
Polyclonal | |
C2CD6 | |
amyotrophic lateral sclerosis 2 chromosomal region candidate gene 11 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151254 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
Q53TS8 | |
C2CD6 | |
KLH conjugated synthetic peptide between 152-181 amino acids from the N-terminal region of human AL2SA | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction