Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Alas1, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP154668

Catalog No. NBP154668

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Alas1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
ALAS1
Synthetic peptides corresponding to ALAS1(aminolevulinate, delta-, synthase 1) The peptide sequence was selected from the N terminal of ALAS1 (NP_000679). Peptide sequence ETSAGPSVVSVKTDGGDPSGLLKNFQDIMQKQRPERVSHLLQDNLPKSVS.
70 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Reconstitute in 50μL distilled water to a final antibody concentration of 1mg/mL.
Western Blot
Western Blot 0.2-1 ug/ml
P13196
5-aminolevulinic acid synthase 1, ALAS, ALAS3, ALASH, ALAS-H, aminolevulinate, delta-, synthase 1, Delta-ALA synthase 1,5-aminolevulinate synthase, nonspecific, mitochondrial, Delta-aminolevulinate synthase 1, EC 2.3.1, EC 2.3.1.37, MIG4, migration-inducing protein 4
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
211
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only