Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Albumin, Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ 1267051ML
Description
Specifically detects Albumin Clone: in Chicken samples, and it is validated for Immunodiffusion, Immunoprecipitation

Specifications
Albumin | |
Polyclonal | |
In PBS. | |
purified chicken albumin | |
RUO | |
Recognizes the ∽45kDa ovalbumin protein. Antibody Target Gene Symbol: ALB Target Synonym: ALB1, ALBUMIN, Albumin 1, Albumin isomer 1, Albumin isomer 2, Albumin isomer 3, Albza, DKFZp779N1935, PRO0883, PRO0903, PRO1341, SA, SERUM ALBUMIN, SERUM ALBUMIN CHAIN A, Serum albumin precursor Entrez Gene Name: albumin | |
−20°C | |
IgG |
Immunodiffusion, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
1 mL | |
Primary | |
Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction