Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin Sheep anti-Rat, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA129266

Catalog No. PA129266


Add to cart

Description

Description

PA1-29266 detects Rat Albumin from rat samples. PA1-29266 has been successfully used in Western blot and ELISA applications. The PA1-29266 immunogen is highly purified normal rat albumin in CFA.

Specifications

Specifications

Rat Albumin
Polyclonal
Unconjugated
Alb
Liquid
ALB
highly purified normal rat albumin in CFA
Purified
RUO
Antibody
Polyclonal
Rat
ELISA, Western Blot
1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
P02770
Alb1, Albza
Sheep
IgG
1 mg
4° C
Primary
24186
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit