Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin Sheep anti-Bovine, Rat, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA185142

Catalog No. PA185142


Add to cart

Description

Description

Sheep anti Rat albumin antibody recognizes rat albumin, a soluble protein which constitutes approximately 50% of blood serum protein, acting as a regulator of blood colloidal osmotic pressure and hence blood volume.

Albumin is a soluble protein which constitutes approximately 50% of blood serum protein, and acts as a regulator of blood colloidal osmotic pressure and blood volume. Albumin is also an important transporter of substances within the bloodstream, such as fatty acids, thyroid hormones, metal ions and steroids.
Specifications

Specifications

Rat Albumin
Polyclonal
Unconjugated
Alb
Liquid
ALB
Native rat albumin protein.
Protein G
RUO
Antibody
Polyclonal
Bovine, Rat
ELISA
5 mg/mL
PBS with 0.09% sodium azide
P02769, P02770
Alb1, Albza
Sheep
IgG
1 mL
4°C or -20°C if preferred
Primary
24186, 280717
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit