Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol Dehydrogenase 1C Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527398

Catalog No. PIPA527398


Add to cart

Description

Description

PA5-27398 targets Alcohol dehydrogenase 1C in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-27398 immunogen is recombit fragment corresponding to a region within amino acids 181 and 375 of Human Alcohol dehydrogenase 1C. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes class I alcohol dehydrogenase, gamma subunit, which is a member of the alcohol dehydrogenase family. Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. Class I alcohol dehydrogenase, consisting of several homo- and heterodimers of alpha, beta, and gamma subunits, exhibits high activity for ethanol oxidation and plays a major role in ethanol catabolism. Three genes encoding alpha, beta and gamma subunits are tandemly organized in a genomic segment as a gene cluster.
Specifications

Specifications

Alcohol dehydrogenase 1C
Polyclonal
Unconjugated
ADH1C
Liquid
ADH1C
Recombit fragment corresponding to a region within amino acids 181 and 375 of Human Alcohol dehydrogenase 1C
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.48 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P00326
ADH3, ADH, gamma subunit, alcohol dehydrogenase 1C, alcohol dehydrogenase 3 (class I), gamma polypeptide, alcohol dehydrogenase subunit gamma, aldehyde reductase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
126
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit