Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALDH3A1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515003

Catalog No. PIPA515003

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Aldehyde dehydrogenases oxidize various aldehydes to the corresponding acids. They are involved in the detoxification of alcohol-derived acetaldehyde and in the metabolism of corticosteroids, biogenic amines, neurotransmitters, and lipid peroxidation. ALDH3A1 forms a cytoplasmic homodimer that preferentially oxidizes aromatic and medium-chain (6 carbons or more) saturated and unsaturated aldehyde substrates. The enzyme is thought to promote resistance to UV and 4-hydroxy-2-nonenal-induced oxidative damage in the cornea.
Specifications

Specifications

ALDH3A1
Polyclonal
PBS with 0.09% sodium azide
P30838
ALDHIII; Aldehyde dehydrogenase family 3 member A1; Aldehyde dehydrogenase 3; Aldehyde dehydrogenase, dimeric NADP-preferring; ALDH3A1; ALDH3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
218
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
ALDH3A1
Liquid
ALDH3A1
KLH conjugated synthetic peptide between 69-99 amino acids from the N-terminal region of human ALDH3A1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit