Learn More
ALDH3B2 Mouse anti-Human, Clone: 3E6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518375
Catalog No. PIMA518375
Add to cart
Description
Description
Peptide Sequence: MKDEPRSTNL FMKLDSVFIW KEPFGLVLII APWNYPLNLT LVLLVGALAA GSCVVLKPSE ISQGTEKVLA EVLPQYLDQS CFAVVLGGPQ ETGQLLEHKL DYIFFTGSPR VGKIVMTAAT KHLTPVTLEL GGKNPCYVDD NCDPQTVANR VAWFCYFNAG QTCVAPDYVL CSPEMQERLL PALQSTITRF YGDDPQSSPN LGRIINQKQF QRLRALLGCG RVAIGGQSNE SDRYIAPTVL VDVQETEPVM QEEIFGPILP IVNVQSVDEA IKFINWQEKP LALYAFSNSS QVVNQMLERT SSGSFGGNEG FTYISLLSVP FGGVGHSGMG RYHGKFTFDT FSHHRTCLLA PSGLEKLKEI HYPPYTDWNQ QLLRWGMGSQ SCTLL
This gene encodes a member of the aldehyde dehydrogenase family, a group of isozymes that may play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. The gene of this particular family member is over 10 kb in length. The expression of these transcripts is restricted to the salivary gland among the human tissues examined. Alternate transcriptional splice variants have been characterized.
Specifications
Specifications
ALDH3B2
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P48448
ALDH8, acetaldehyde dehydrogenase 8, aldehyde dehydrogenase 8, aldehyde dehydrogenase family 3 member B2
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
222
ELISA
3E6
Unconjugated
ALDH3B2
Liquid
ALDH3B2
ALDH3B2 (AAH07685, 1 a.a. ∽ 385 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human