Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG1 Rabbit anti-C. elegans, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA1031X

Catalog No. PIPA1031X

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

A homolog of rde-1 that is involved in RNA interference and affects developmental timing along with alg-2 and dcr-1 by regulating expression of the lin-4 and let-7 small temporal RNAs.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ALG1
Polyclonal
Unconjugated
Argonaute, Argonaute-like
Rabbit
Antigen affinity chromatography
RUO
181504
-20°C
Liquid
Western Blot
1 mg/mL
alg-1
ALG-1
Peptide sequence DLEEIPNSPPTQPQTFSDVPQR
20 μL
Primary
C. elegans
Antibody
IgG
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit