Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALG10 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526960

Catalog No. PIPA526960

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

ALG10 encodes a membrane-associated protein that adds the third glucose residue to the lipid-linked oligosaccharide precursor for N-linked glycosylation. That is, it transfers the terminal glucose from dolichyl phosphate glucose (Dol-P-Glc) onto the lipid-linked oligosaccharide Glc2Man9GlcNAc(2)-PP-Dol. The rat protein homolog was shown to specifically modulate the gating function of the rat neuronal ether-a-go-go (EAG) potassium ion channel.
Specifications

Specifications

ALG10
Polyclonal
PBS with 0.09% sodium azide
Q5BKT4
DIE2, KCR1, alpha-1,2-glucosyltransferase ALG10-A, alpha-2-glucosyltransferase ALG10-A, alpha2-glucosyltransferase, asparagine-linked glycosylation 10 homolog (yeast, alpha-1,2-glucosyltransferase), asparagine-linked glycosylation protein 10 homolog A, derepression of ITR
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84920
Immunohistochemistry, Western Blot
Unconjugated
ALG10
Liquid
ALG10
KLH conjugated synthetic peptide between 17-43 amino acids from the N-terminal region of human ALG10
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit