Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alivin 1 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA146258

Catalog No. PIPA146258

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

This antibody has 81% sequence homology with human.

A novel primary response gene, Alivin 1 (ali1), is an activity-dependent gene and promotes survival of neurons. Sequence analyses reveal that rat, mouse, and human Ali1 proteins contain seven leucine-rich repeats, one IgC2-like loop and a transmembrane domain, and display homology to Kek and Trk families. Results suggest that expression of ali1 promotes depolarization-dependent survival of the granule neuron. Mouse ali1 was mapped to a locus approximately 55. 3 cM from the centromere on chromosome 15 that is syntenic to positional candidate loci for familial Alzheimer′s disease type 5 and Parkinson′s disease 8 on human chromosome 12.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Alivin 1
Polyclonal
Unconjugated
Q80ZD9, Q7TNJ4
AMIGO2
Synthetic peptide mapping to residues 449-464 of mouse and rat Alivin 1.
100 μg
Primary
Mouse, Rat
Antibody
IgG
Western Blot
1 mg/mL
Amigo2
Amphoterin-induced protein 2, AMIGO-2, ALI1, DEGA
Rabbit
Antigen affinity chromatography
RUO
105827, 300186
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit