Learn More
anti-ALK-1, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP132200
Catalog No. NBP132200
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
ALK-1
0.78mg/mL
Western Blot 1:500-1:3000
Affinity Purified
ACVRL1
Recombinant protein encompassing a sequence within the center region of the cytoplasmic domain of human ACVRL1. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Primary
94
Human, Mouse
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRLK1ORW2, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I
Rabbit
IgG
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.