Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Alkaline Phosphatase/ALPL APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems FAB1448A025
This item is not returnable.
View return policy
Description
Alkaline Phosphatase/ALPL Monoclonal specifically detects Alkaline Phosphatase/ALPL in Human samples. It is validated for Flow Cytometry.Specifications
Alkaline Phosphatase/ALPL | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
ALPL | |
Human liver, bone and kidney-derived Alkaline Phosphatase/ALPL | |
25 Tests | |
Primary | |
Detects liver, bone and kidney Alkaline Phosphatase/ALPL from human tissue (2). | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
B4-78 | |
Flow Cytometry 10 uL/10^6 cells | |
Akp2, Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALP, TNSALPMGC167935 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
249 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction