Learn More
SNAP25 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSA00051W
Catalog No. PIOSA00051W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water.
SNAP25 is a t-SNARE involved in the molecular regulation of neurotransmitter release. It may play a role in the synaptic function of specific neuronal systems. SNAP25 associates with proteins involved in vesicle docking and membrane fusion. It also regulates plasma membrane recycling through its interaction with CENPF. Mutations in the gene can results in myasthenic syndrome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SNAP25
Polyclonal
whole serum with no preservative
P60879
SNAP25
A synthetic peptide from C-terminal region of mouse Alstrom syndrome protein 1 homolog
RUO
20614
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunohistochemistry, Western Blot
Unconjugated
Snap25
ALMS1
Rabbit
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions
Product Suggestions