Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALOX15B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526208

Catalog No. PIPA526208

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ALOX15B encodes a member of the lipoxygenase family of structurally related nonheme iron dioxygenases involved in the production of fatty acid hydroperoxides. The encoded protein converts arachidonic acid exclusively to 15S-hydroperoxyeicosatetraenoic acid, while metabolizing linoleic acid less effectively. This gene is located in a cluster of related genes and a pseudogene that spans approximately 100 kilobases on the short arm of chromosome 17.
Specifications

Specifications

ALOX15B
Polyclonal
PBS with 0.09% sodium azide
O15296
15-LOX-2, 15-LOX-B, 15S-lipoxygenase, arachidonate 15-lipoxygenase 2, arachidonate 15-lipoxygenase B, arachidonate 15-lipoxygenase type II, arachidonate 15-lipoxygenase, second type, arachidonate omega(6) lipoxygenase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
247
Immunohistochemistry, Western Blot
Unconjugated
ALOX15B
Liquid
ALOX15B
KLH conjugated synthetic peptide between 597-627 amino acids from the C-terminal region of human ALOX15B
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit