Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLAP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515067

Catalog No. PIPA515067

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse, porcine, non-human primate, rat and rabbit based on sequence homology.

ALOX5AP is a protein which, with 5-lipoxygenase, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.
Specifications

Specifications

FLAP
Polyclonal
PBS with 0.09% sodium azide
P20292
Arachidonate 5-lipoxygenase-activating protein; FLAP; MK-886-binding protein; ALOX5AP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
241
Immunohistochemistry, Western Blot
Unconjugated
ALOX5AP
Liquid
ALOX5AP
KLH conjugated synthetic peptide between 86-117 amino acids from the central region of human ALOX5AP
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit