Learn More
ORM1 Rabbit anti-Human, HRP, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA127247
Catalog No. PA127247
Add to cart
Description
Description
PA1-27247 detects alpha 1 Acid Glycoprotein from human samples. PA1-27247 has been successfully used in immunohistochemistry, electron microscopy, and Western blot procedures. The PA1-27247 immunogen is alpha 1 Acid Glycoprotein from human plasma.
This gene encodes a key acute phase plasma protein. Because of its increase due to acute inflammation, this protein is classified as an acute-phase reactant. The specific function of this protein has not yet been determined; however, it may be involved in aspects of immunosuppression.
Specifications
Specifications
ORM1
Polyclonal
HRP
ORM1
Liquid
ORM1
Alpha 1 Acid Glycoprotein from human plasma.
IgG fraction
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Immunomicroscopy, Western Blot
10 mg/mL
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2
P02763
AGP-A, AGP1, ORM
Rabbit
IgG
1 mg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5004