Learn More
SERPINA3 Mouse anti-Human, Clone: 1AC10-109.5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110629
Catalog No. 01-671-582
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10629 detects alpha-1 Antichymotrypsin from human samples. MA1-10629 has been successfully used in ELISA applications. The MA1-10629 immunogen is purified human serum alpha-1 Antichymotrypsin.
The protein encoded by this gene is a plasma protease inhibitor and member of the serine protease inhibitor class. Polymorphisms in this protein appear to be tissue specific and influence protease targeting. Variations in this proteiNULLs sequence have been implicated in Alzheimer′s disease, and deficiency of this protein has been associated with liver disease. Mutations have been identified in patients with Parkinson disease and chronic obstructive pulmonary disease.
Specifications
Specifications
SERPINA3
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
P01011
AACT, ACT, GIG24, GIG25, MGC88254
Mouse
IgG2a
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
12
ELISA
1AC10-109.5
Unconjugated
SERPINA3
Liquid
SERPINA3
Purified human serum alpha-1 Antichymotrypsin.
Protein G
RUO
Antibody
Monoclonal
Human