Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-1 Antitrypsin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527311

Catalog No. PIPA527311


Add to cart

Description

Description

PA5-27311 targets alpha 1 Antitrypsin in WB applications and shows reactivity with Human samples. The PA5-27311 immunogen is recombit fragment corresponding to a region within amino acids 192 and 418 of Human alpha 1 Antitrypsin. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is secreted and is a serine protease inhibitor whose targets include elastase, plasmin, thrombin, trypsin, chymotrypsin, and plasminogen activator. Defects in this gene can cause emphysema or liver disease. Several transcript variants encoding the same protein have been found for this gene.
Specifications

Specifications

alpha-1 Antitrypsin
Polyclonal
Unconjugated
SERPINA1
Liquid
SERPINA1
Recombit fragment corresponding to a region within amino acids 192 and 418 of Human alpha 1 Antitrypsin
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.35 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P01009
A1A, A1AT, AAT, PI, PI1, PRO2275, alpha1AT, alpha-1 protease inhibitor, alpha-1-antiproteinase, alpha-1-antitrypsin, alpha-1-antitrypsin null, protease inhibitor 1 (anti-elastase), alpha-1-antitrypsin, serine (or cysteine) proteinase inhibitor, clade A, member 1, serpin A1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5265
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit