Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A2M Chicken anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA129889

Catalog No. PIPA129889


Add to cart

Description

Description

PA1-29889 detects alpha 2 Macroglobulin from rat, human, mouse samples. PA1-29889 has been successfully used in Western blot & ELISA applications. The PA1-29889 immunogen is full length protein (Human). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Alpha 2-Macroglobulin (a2M), is a ~720-kDa homotetrameric glycoprotein composed of four identical ~180 kDa subunits. aAlpha 2M shares with other alpha-macroglobulins, like the complement components C3 and C4 and the pregcy zone protein PZP, an extraordinary binding capacity for a variety of ligands. This allows the alpha-macroglobulins to serve as humoral defense barriers against foreign peptides in the plasma. Alpha 2M interacts and captures virtually any proteinase, often referred to as a panprotease inhibitor. In the brain of Alzheimer ft.s disease (AD) patients, Alpha 2M also has been localized to diffuse amyloid plaques, supporting an important role for alpha 2M in AD etiopathology.
Specifications

Specifications

A2M
Polyclonal
Unconjugated
A2M
Liquid
A2M
Full length protein (Human).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P01023, Q6GQT1, P06238
CPAMD5, DKFZp779B086, FWP007, S863-7
Chicken
IgY
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2, 232345, 24153
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit