Learn More
alpha-2a Adrenergic Receptor Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA130159
Catalog No. PIPA130159
Add to cart
Description
Description
PA1-30159 detects alpha 2a Adrenergic receptor from mouse, rat, and human samples. PA1-30159 has been successfully used in immunohistochemistry (frozen tissue) applications. The PA1-30159 immunogen is peptide - (C)KASRWRGRGNREKR (Cysteine added for conjugation) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
Specifications
alpha-2a Adrenergic Receptor
Polyclonal
Unconjugated
ADRA2A
Liquid
ADRA2A
Peptide - (C)KASRWRGRGNREKR (Cysteine added for conjugation)
IgG fraction
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
0.47 mg/mL
Antibody with no preservative
P08913, Q01338, P22909
ADRA2, ADRA2R, ADRAR, ALPHA2AAR, ZNF32
Rabbit
IgG
50 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11551, 150, 25083