Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-alpha-Actin Clone: 1A4,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 113200500UL
Description
Specifically detects α-Actin Clone: 1A4 in Baboon, Human, Rabbit samples, and it is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Actin is a cytoskeletal protein. Smooth muscle actin forms smaller, less ordered contractile units than actin from skeletal or cardiac muscle. The activity and state of assembly of smooth muscle actin is controlled by calcium-regulated phosphorylation of one of the myosin light chains.Specifications
α-Actin | |
Monoclonal | |
Unconjugated | |
Mouse | |
500 μL | |
Primary | |
Baboon, Human, Rabbit | |
Supernatant |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
1A4 | |
In 150mM NaCl, 20mM Tris-borate, BHC-6 carrier polysaccharides, pH 7.5. | |
a synthetic peptide corresponding to amino acids 1-10 of human smooth muscle α-actin. | |
RUO | |
Recognizes α-actin in smooth muscle cells of blood vessels and parenchymal tissue of the intestine, testis, and ovary. Does not recognize actin from fibroblasts (β- and γ-cytoplasmic), striated muscle (α-sarcomeric), or myocardium (α-myocardial). Antibody Target Gene Symbol: ACTA2 Target Synonym: 0610041G09RIK, a-SMA, AAT6, ACTSA, Actvs, ALPHA ACTIN VSM, ALPHA SMA, ALPHA SMOOTH MUSCLE ACTIN, ALPHA-ACTIN 2, FLJ45296, MGC127800, RGD: 621676, Similar to vascular alpha actin, Sm alpha actin, SMalphaA, SMOOTH MUSCLE ALPHA ACTIN Entrez Gene Name: actin, alpha 2, smooth muscle, aorta | |
2°C to 8°C | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction