Learn More
alpha Actinin 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA527862
Description
PA5-27862 targets alpha Actinin 3 in WB applications and shows reactivity with Human, mouse, and Rat samples. The PA5-27862 immunogen is recombit fragment corresponding to a region within amino acids 1 and 363 of Human alpha Actinin 3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Alpha-actinin is an actin-binding protein with multiple roles in different cell types. This gene expression is limited to skeletal muscle. It is localized to the Z-disc and analogous dense bodies, where it helps to anchor the myofibrillar actin filamentsSpecifications
alpha Actinin 3 | |
Polyclonal | |
Unconjugated | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q08043, O88990 | |
11474, 171009, 89 | |
Rabbit | |
IgG | |
Primary | |
100 μL | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Antibody |
Western Blot | |
1 mg/mL | |
Liquid | |
ACTN3 | |
F-actin cross-linking protein, alpha-actinin skeletal muscle, alpha-actinin-3 | |
ACTN3 | |
Recombit fragment corresponding to a region within amino acids 1 and 363 of Human alpha Actinin 3 | |
Polyclonal | |
Antigen affinity chromatography | |
RUO | |
Human,Mouse,Rat |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.