Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Actinin 4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522259

Catalog No. PIPA522259


Add to cart

Description

Description

PA5-22259 targets alpha Actinin 4 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human, Rat and mouse samples. The PA5-22259 immunogen is a recombit fragment corresponding to a region within amino acids 206 and 542 of alpha Actinin 4 (Uniprot IDNo.O43707). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

alpha Actinin 4
Polyclonal
Unconjugated
ACTN4
Liquid
ACTN4
Recombit fragment corresponding to a region within amino acids 206 and 542 of alpha Actinin 4 (Uniprot IDNo.O43707)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1.03 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O43707, P57780, Q9QXQ0
ACTININ-4, FSGS, FSGS1, F-actin cross-linking protein, actinin alpha4 isoform, alpha-actinin-4, focal segmental glomerulosclerosis 1, non-muscle alpha-actinin 4
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
60595, 63836, 81
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit