Learn More
alpha Actinin 1 Rabbit anti-Chicken, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA128036
Catalog No. PA128036
Add to cart
Description
Description
PA1-28036 detects alpha Actinin from chicken samples. Expected to cross-react with a wide range of tther species due to sequence homology. PA1-28036 has been successfully used in immunofluorescence and Western blot applications. The PA1-28036 immunogen is full length native alpha actinin purified from chicken gizzard. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Alpha Actinin is an actin binding protein present in both muscle and non-muscle cells. It has a molecular weight of 100,000 daltons and forms dimers in solution. In normal skeletal muscle alpha actinin is associated with the z-discs that define muscle sarcomeres. In smooth muscle alpha actinin has been detected in dense bodies and plaques characteristic of the tissue. Immunofluorescent labeling of a large variety of cells using antibody to alpha actinin reveals extensive association of the proteins with actin containing stress fibers and in particular with their membrane bound termini.
Specifications
Specifications
alpha Actinin 1
Polyclonal
whole serum with 15mM sodium azide
P05094
RCJMB04_23c5, ACTN1
Rabbit
IgG
RUO
Antibody
Polyclonal
Chicken
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
ACTN1
Liquid
ACTN1
Full length native alpha actinin purified from chicken gizzard.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
373918