Learn More
alpha Amylase 1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA129115
Catalog No. PA129115
Add to cart
Description
Description
PA1-29115 detects alpha Amylase from human samples. PA1-29115 has been successfully used in immunohistochemistry (frozen tissue), immunohistochemistry (paraffin tissue), Western blot and ELISA applications. Positive control of pancreas suggested. The PA1-29115 immunogen is purified human salivary a-amylase.
Specifications
Specifications
alpha Amylase 1
Polyclonal
PBS with no preservative; pH 7.2
P04745
AMY1A
Purified human alpha-amylase.
500 μL
4° C, do not freeze
Primary
276
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
AMY1A
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal
Human