alpha B Crystallin Polyclonal Antibody, Invitrogen™

alpha B Crystallin Antibody, PA1-16951, from Invitrogen™. Species Reactivity: Human; Applications: Immunocytochemistry, Immunofluorescence, Western Blot.

Manufacturer: Invitrogen PA116951

Catalog No. 01-950-180

  Check Availability
Add to Cart

    Specifications

    Formulation 100μL purified antibody (0.9mg/mL) in tris citrate and 0.1% sodium azide; pH 7-8
    Gene Accession No. P02511
    Applications Immunocytochemistry,Immunofluorescence,Western Blot
    Gene ID (Entrez) 1410
    Cross Reactivity Human
    Gene Alias Alpha crystallin B chain antibody, CryAB antibody, Alpha (B)-crystallin antibody, Rosenthal fiber componet antibody, Heat-shock protein beta-5 antibod
    Gene Symbols CRYAB
    Isotype IgG
    Species Reactivity Human, Mouse
    Storage Requirements Store at 4° C
    Antigen alpha B Crystallin
    Format Purified
    Primary or Secondary Primary
    Monoclonal or Polyclonal Polyclonal
    Conjugate Unlabeled
    Host Species Rabbit
    Immunogen Synthetic peptide between amino acids 100-175 of the human Crystallin AB protein.
    Quantity 100μL

    alpha B Crystallin Antibody, PA1-16951, from Invitrogen. Species Reactivity: Human; Applications: Immunocytochemistry, Immunofluorescence, Western Blot.

    Crystallin AB (CryAB) is a developmentally regulated small heat shock protein known to protect cells from stress damage. It may also play an important role in the Golgi reorganization during cell division. CryAB is known to be associated with a number of neurodegenerative pathologies such as Alzheimer disease amd multiple sclerosis. As well, it has been found to be expressed in breast tumors of patients with shorter survival, suggesting that it may be a useful marker in identifying aggressive breast cancer.
    Host Species: Rabbit
    Cross Reactivity: Human
    Isotype: IgG
    Immunogen: Synthetic peptide between amino acids 100-175 of the human Crystallin AB protein.
    Formulation: 100µL purified antibody (0.9mg/mL) in tris citrate and 0.1% sodium azide; pH 7-8
    Synonyms: Alpha crystallin B chain antibody, CryAB antibody, Alpha (B)-crystallin antibody, Rosenthal fiber componet antibody, Heat-shock protein beta-5 antibod
    Uniprot Accession: P02511
    Entrez GeneID: 1410
    Gene Symbol: CRYAB
    This antibody is expected to react with rat, ovine, rabbit, monkey, hamster, bovine and porcine based on sequence homology.

    • Purchase any three Invitrogen™ Antibodies and receive a fourth antibody at no additional cost. To take advantage of this offer, apply for the NLSU program.

      New Offer Buy Now

      Expires 2018-06-30 Redemption Only!

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit