Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha/beta Tubulin, clone: TU-08, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA119399

Catalog No. 01-676-147

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Tubulin is a heterodimer consisting of alpha and beta-tubulin subunits. It is nearly ubiquitous among eukaryotic cells. The alpha and beta tubulins, which are each about 55kDa MW, are homologous but not identical. Alpha-beta tubulin heterodimer is the basic building block of microtubules. This intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis

Specifications

Specifications

alpha Tubulin + beta Tubulin
Monoclonal
1mg/mL
Assay dependent
TUBA1A
Purified
TUBA1A, TUBB, TUBB1
Microtubule proteins from porcine brain.
Purified
RUO
Antibody
Monoclonal
Human, Murine, Porcine
Immunocytochemistry, Western Blot
TU-08
Unconjugated
TBS with 15mM sodium azide; pH 8.0
P07437, Q71U36, Q9H4B7, A2AQ07, P68369, P02550, Q767L7
TUBA1A, TUBB1, FLJ30169, H2 ALPHA, Testis specific alpha tubulin, TUBA1, Tubulin alpha
Murine
IgM
100μg
Store at 4°C
Primary
203068, 7846, 81027
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit