Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Dystroglycan Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA528179
Description
Recommended positive controls: U87-MG, U87-MG membrane fraction extract. Predicted reactivity: Mouse (92%), Rat (92%), Dog (92%), Cat (95%), Rabbit (94%), Rhesus Monkey (98%), Bovine (93%).
alpha Dystroglycan Polyclonal antibody specifically detects alpha Dystroglycan in Equine, Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
alpha Dystroglycan | |
Polyclonal | |
Unconjugated | |
DAG1 | |
156DAG; A3a; AGRNR; Alpha-DG; Alpha-dystroglycan; Beta-DG; Beta-dystroglycan; D9Wsu13e; DAG; DAG1; Dag-1; DG; Dp427; Dp71; dystroglycan; dystroglycan 1; dystroglycan 1 (dystrophin-associated glycoprotein 1); dystrophin associated glycoprotein 1; Dystrophin-associated glycoprotein 1; MDDGA9; MDDGC7; MDDGC9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100053150, 13138, 1605 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.21 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q14118, Q62165 | |
DAG1 | |
Recombinant fragment corresponding to a region within amino acids 471 and 752 of Human alpha Dystroglycan | |
100 μL | |
Primary | |
Equine, Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction