Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anti-alpha Gal Polyclonal Antibody (Chicken) , TCI America™

Anti-alpha Gal Polyclonal Antibody (Chicken), TCI America™
Supplier: TCI America A31231VIAL
Specifications
| αGal | |
| Polyclonal | |
| 1 Vial | |
| Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Chicken | |
| Primary | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction