Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Galactosidase Chicken anti-Human, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA129518

Catalog No. PIPA129518


Add to cart

Description

Description

PA1-29518 detects Galactosidase alpha from human samples. PA1-29518 has been successfully used in ELISA and Western blot applications. The PA1-29518 immunogen is: Synthetic peptide- amino acids 396-407 and 55-64 of Human Galactosidase alpha. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

alpha Galactosidase
Polyclonal
Unconjugated
GLA
Liquid
Chicken
IgY
250 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
2717
ELISA, Western Blot
1 mg/mL
PBS with 40% glycerol and no preservative; pH 7.4
P06280
GLA
Synthetic peptide- amino acids 396-407 and 55-64 of Human Galactosidase alpha.
Ig fraction
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit