Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-MSH Rabbit anti-Porcine, Rat, Polyclonal, Invitrogen

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA136171

Catalog No. 01-672-120


Add to cart

Description

Description

A suggested positive control for IHC is Bouin-fixed paraffin or Stefanini-fixed frozen sections of pig pituitary. Over-night incubation at 4°C is recommended. Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. For long-term storage, aliquot and store at -20°C.

The melanocortins are regulatory neuropeptidesderived by post-translational processing of the larger 30 kDa, pro-opiomelanocortin (POMC) precursor, and include the adrenocorticotropic hormone (ACTH) and the melanocyte stimulating hormones (?-, ?-, and ?-MSH). The melanocortins are produced primarily in the anterior and intermediate lobes of the pituitary, in the arcuate nucleus of the hypothalamus, and to a lower level in various other peripheral tissues. In addition to their well established melanotropic and adrenocorticotropic actions, these peptides have been reported to exert a broad variety of physiological actions including those related to central neural function such as behavior, memory, cognition, and regulation of feeding. Peripherally, melanocortins have immunomodulatory and neurotrophic properties.
Specifications

Specifications

alpha-MSH
Polyclonal
whole serum with 0.1% sodium azide
P01192, P01194
ACTH, CLIP, LPH, MSH, NPP, POC
Rabbit
IgG
RUO
Antibody
Polyclonal
Porcine, Rat
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
Pomc
Lyophilized
POMC
Synthetic alpha-MSH conjugated to BSA
50 μL
4° C
Primary
24664, 396863
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit