Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAK2 Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137067

Catalog No. 01-673-046

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and mouse based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is breast carcinoma.

Alpha PAK p68, beta PAK p65 and gamma PAK p62 are three isoforms of serine/threonine kinases. These three isoforms complex specifically with Rac1 and Cdc42 in their active GTP-bound state, inhibiting their intrinsic GTPase activity leading to their autophosphorylation.
Specifications

Specifications

PAK2
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.4
Q13177, Q64303
PAK65, PAKgamma, PAK-2, S6/H4 kinase, gamma-PAK, p21 CDKN1A-activated kinase 2, p21-activated kinase 2, p58, serine/threonine-protein kinase PAK 2
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
29432, 5062
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PAK2
Liquid
PAK2
Synthetic peptide corresponding to aa 525-545 of rat alpha PAK
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit