Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Synuclein Mouse anti-Human, Clone: Syn 211, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112874

Catalog No. 01-674-078


Add to cart

Description

Description

A suggested positive control for this product is SHSY-5Y cells. It will not cross-react with mouse or rat.

Alpha-Synuclein belongs to the Synuclein family, which includes beta and gamma Synuclein, and is predomitly expressed in neurons, and concentrated at synaptic terminals. Synucleins may be involved in the regulation of dopamine release and transport and the modulation of synaptic vesicle function. Alpha-Synuclein is a small, presynaptic protein that is the major nonamyloid component of the pathological inclusions characteristic of a wide range of neurodegenerative disorders, collectively known as synucleinopathies. Alpha-Synuclein is involved in the formation of SNARE complexes, and most significantly, aggregated alpha-synuclein is one of the major components found in the Lewy bodies that occur in Parkinson ft.s disease (PD) and other neurodegenerative disorders. Mutation of the alpha-synuclein gene is associated with familial forms of PD. Alpha-Synuclein is known to reduce the fibrillization of the microtubule-associated protein, tau, to bind various targets such as 14-3-3, protein kinase C, synphilin-1, Elk and Tat-binding protein 1. The alpha and beta synucleins are found primarily in brain tissue where they are seen mainly in neuronal cytosol and presynaptic terminals. The antibody specific for alpha-synuclein does not cross react with beta-synuclein and will be useful for the confirmation of Parkinson ft.s disease and Lewy body dementia and other studies for the localization and detection of alpha-synuclein. Four alternatively spliced transcripts that encode two different isoforms of the alpha-synuclein protein have been identified.
Specifications

Specifications

alpha Synuclein
Monoclonal
0.5 mg/mL
PBS with 0.1% BSA and 0.1% sodium azide; pH 7.2
P37840
Non-A beta component of AD amyloid
Mouse
IgG1
100 μg
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
6622
ELISA, Immunohistochemistry (Paraffin), Western Blot
Syn 211
Unconjugated
SNCA
Liquid
SNCA
Recombit human alpha-synuclein
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit