Learn More
alpha Synuclein Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA138701
Description
Synuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's PD) and breast cancer. In AD, a peptide derived from alpha synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of alpha synuclein has been linked to many familial cases of PD. In breast cancer increased expression of gamma synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.
This antibody is predicted to react with bovine, chicken, canine, mouse, porcine, rabbit and rat based on sequence homology.A suggested positive control is brain (posterior cingulated gyrus) tissue.
Specifications
alpha Synuclein | |
Polyclonal | |
SNCA | |
NACP, PARK1, PARK4, PD1, alpha-synuclein, non A-beta component of AD amyloid, synuclein alpha-140 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6622 | |
4° C, do not freeze | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
P37840 | |
SNCA | |
Synthetic peptide near C-terminus of human synuclein-alpha | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.