Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Tubulin, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522258

Catalog No. PIPA522258

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA5-22258 targets alpha Tubulin 4a in ICC, IF, IHC (P), and WB applications and shows reactivity with Human and mouse samples.The PA5-22258 immunogen is a recombinant fragment corresponding to a region within amino acids 21 and 243 of alpha Tubulin 4a (Uniprot ID#P68366).
Specifications

Specifications

alpha Tubulin 4a
Polyclonal
Unconjugated
TUBA4A
Purified
TUBA4A
Recombinant fragment corresponding to a region within amino acids 21 and 243 of alpha Tubulin 4a (Uniprot ID#P68366)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.92mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P68366, P68368
H2-ALPHA, TUBA1, tubulin H2-alpha, tubulin alpha-1 chain, tubulin alpha-4A chain, tubulin, alpha 1 (testis specific)
Rabbit
IgG
100μL
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
7277
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit