Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TUBA1/3/4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539836
Description
Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa MW, are homologous but not identical. Alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predomitly found in morphologically differentiated neurologic cells.Specifications
alpha Tubulin | |
Polyclonal | |
Unconjugated | |
TUBA4A | |
TUBA1/3/4, H2-ALPHA, TUBA1, tubulin H2-alpha, tubulin alpha-1 chain, tubulin alpha-4A chain, tubulin, alpha 1, B-ALPHA-1, LIS3, TUBA3, alpha-tubulin 3, hum-a-tub1, hum-a-tub2, tubulin B-alpha-1, tubulin alpha-1A chain, tubulin alpha-3 chain, tubulin, alpha 3, tubulin, alpha, brain-specific | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10376, 112714, 22142, 22143, 22145, 22146, 300218, 316531, 500929, 64158, 7277, 7278, 7846, 84790 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P68363, P68366, Q13748, Q6PEY2, Q71U36, Q9BQE3, P05213, P68368, P68369, P68373, P68370, Q5XIF6, Q6AYZ1, Q6P9V9 | |
TUBA1A, TUBA1B, TUBA1C, TUBA3C, TUBA3E, TUBA4A | |
A synthetic non-phosphopeptide derived from human TUBA1/3/4 around the phosphorylation site of Tyr272 (A-T-YP-A-P) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction