Learn More
Alpha4 Mouse anti-Human, Mouse, Clone: 5F6, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05930
Catalog No. 05-930-MI
Add to cart
Description
Description
This Anti-Alpha4 Antibody, clone 5F6 is validated for use in western blotting for the detection of Alpha4.
Specifications
Specifications
Alpha4
5F6
NP_001542.1
Murine
IgG1
200μL
Primary
Human, Murine
Western Blot
Unconjugated
Supernatant
N-terminal 6His-tagged fusion protein corresponding to full length mouse Alpha4
Mouse culture supernatant
−20°C for 24 months.
Monoclonal