Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA527667
Description
Recommended positive controls: THP-1. Predicted reactivity: Pig (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ALS Polyclonal antibody specifically detects ALS in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
ALS | |
Polyclonal | |
Unconjugated | |
IGFALS | |
ACLSD; Albs; Als; IGFALS; insulin like growth factor binding protein acid labile subunit; insulin-like growth factor binding protein complex acid labile chain; insulin-like growth factor binding protein complex acid-labile subunit; insulin-like growth factor binding protein, acid labile subunit; insulin-like growth factor-binding protein complex acid labile chain; insulin-like growth factor-binding protein complex acid labile subunit; mKIAA4111 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3483 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P35858 | |
IGFALS | |
Recombinant fragment corresponding to a region within amino acids 375 and 605 of Human ALS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction