Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALY Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA522314
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, Mouse brain. Predicted reactivity: Mouse (96%), Rat (98%), Chicken (82%), Rhesus Monkey (98%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ALY Polyclonal antibody specifically detects ALY in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
ALY | |
Polyclonal | |
Unconjugated | |
ALYREF | |
ally of AML-1 and LEF-1; ALY; ALY/REF; Aly/REF export factor; Aly/REF export factor L homeolog; aly/REF export factor-A; alyref; alyref.L; alyref-a; BEF; bZIP enhancing factor; bZIP-enhancing factor BEF; hypothetical protein LOC537706; REF; Ref1; REF1-I; Refbp1; RNA and export factor binding protein 1; RNA and export factor-binding protein 1; Tcra enhancer-binding factor interacting protein 1; THO complex 4; THO complex subunit 4; THO complex subunit 4-A; tho4; Tho4-A; THOC4; thoc4-a; transcriptional coactivator Aly/REF; XELAEV_18043750mg; zgc:171753 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10189, 21681 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.63 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O08583, Q86V81 | |
ALYREF | |
Recombinant fragment corresponding to a region within amino acids 1 and 233 of ALY (Uniprot ID#Q86V81) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction