Learn More
anti-Alzheimer Precursor Protein A4, aa 66-81, N-terminus, Clone: 22C11, Millipore™ (Chemicon™)
Manufacturer:  Millipore Sigma MAB348100UL/DEL
 View more versions of this product
Catalog No. 50-175-303
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Alzheimer Precursor Protein A4, aa 66-81, N-terminus
Ascites
IgG1
RUO
Primary
Fish,Human,Monkey,Murine,Porcine,Rat
Unlabeled
Murine
100μL
Neuroscience
Monoclonal