Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMBRA1 Rabbit anti-Bovine, Chicken, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523212

Catalog No. PIPA523212


Add to cart

Description

Description

Ambra1, a WD40 domain containing protein, is a novel activating molecule in Beclin 1-regulated autophagy and plays a role in the development of the nervous system. The protein binds to Beclin 1 and favors Beclin 1/Vps34 interaction. The Ambra1 gene localized on human chromosome 11p11.2, encodes the protein that is highly conserved in vertebrates only and its expression is mostly confined to the neuroepithelium during early neurogenesis.
Specifications

Specifications

Ambra1
Polyclonal
Unconjugated
AMBRA1
Liquid
AMBRA1
Synthetic peptide (24 GAQRLLQELVEDKTR 38) of Ambra1 protein.
Protein G
RUO
Antibody
Polyclonal
Bovine, Chicken, Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9C0C7, A2AH22
DCAF3, WDR94, DDB1 and CUL4 associated factor 3, WD repeat domain 94, activating molecule in BECN1-regulated autophagy protein 1, activating molecule in beclin-1-regulated autophagy
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
228361, 423194, 517263, 55626, 59319
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit