Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

amelogenin (amelogenesis imperfecta 1, X-linked), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human AMELX protein.

Manufacturer:  Abnova Corporation H00000265B01

Catalog No. 89-004-889


Add to cart

Description

Description

This gene encodes a member of the amelogenin family of extracellular matrix proteins. Amelogenins are involved in biomineralization during tooth enamel development. Mutations in this gene cause X-linked amelogenesis imperfecta. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq

Sequence: MGTWILFACLLGAAFAMPLPPHPGHPGYINFSYEVLTPLKWYQSIRPPYPSYGYEPMGGWLHHQIIPVLSQQHPPTHTLQPHHHIPVVPAQQPVIPQQPMMPVPGQHSMTPIQHHQPNLPPPAQQPYQPQPVQPQPHQPMQPQPPVHPMQPLPPQPPLPPMFPMQPLPPMLPDLTLEAWPSTDKTKREEVD
Specifications

Specifications

amelogenin (amelogenesis imperfecta 1, X-linked)
Unconjugated
No additive
NM_001142.2
AIH1|ALGN|AMG|AMGL|AMGX
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
265
Western Blot
Mouse polyclonal antibody raised against a full-length human AMELX protein.
AMELX
Serum
AMELX
AMELX (NP_001133.1, 1 a.a. ~ 191 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit