Learn More
AMH Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA526938
Catalog No. PIPA526938
Add to cart
Description
Description
Anti mullerian hormone (AMH) is a member of the TGF beta superfamily. It is secreted as a homodimeric 140kD disulphide linked precursor that is cleaved to release the mature 30kD homodimer. Originally classified as a foetal testicular hormone that inhibits Mullerian duct development, AMH is expressed post natally by immature Sertoli cells, and to a lesser degree by granulosa cells. AMH plays a role in testicular differentiation and in the regulation of ovarian follicle growth.
Specifications
Specifications
AMH
Polyclonal
Unconjugated
AMH
Liquid
AMH
KLH conjugated synthetic peptide between 424-451 amino acids from the central region of human AMH
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P03971, P27106
MIF, MIS, Mullerian inhibiting factor, Mullerian inhibiting substance, anti-Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance
Rabbit
IgG
200μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11705, 268